Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arjunic acid (Arjuntriterpenic acid) | 31298-06-3 | 5 MG
SDP

Catalog No. 5000421203
Encompass_Preferred

Arjunic acid (Arjuntriterpenic acid) is a triterpenoid compound isolated from *Sanguisorba officinalis L*. It is intended for research use only and is not sold to patients.

  • High purity of 99.19%
  • Molecular weight: 488.70
  • Chemical formula: C30H48O5
  • Appearance: Solid, white to off-white
  • Classified as a triterpene
  • Isolated from *Sanguisorba officinalis Linn.*
  • Recommended storage at 4°C, sealed and away from moisture
  • Soluble in DMSO at 50 mg/mL (102.31 mM) with ultrasonic assistance

Catalog No. 50-004-21203 Supplier Medchemexpress LLC Supplier No. HYN28955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Arjunic acid (Arjuntriterpenic acid) is a triterpenoid compound isolated from *Sanguisorba officinalis L*. It is intended for research use only and is not sold to patients.

  • High purity of 99.19%
  • Molecular weight: 488.70
  • Chemical formula: C30H48O5
  • Appearance: Solid, white to off-white
  • Classified as a triterpene
  • Isolated from *Sanguisorba officinalis Linn.*
  • Recommended storage at 4°C, sealed and away from moisture
  • Soluble in DMSO at 50 mg/mL (102.31 mM) with ultrasonic assistance

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.