Learn More
TARGETMOL CHEMICALS INC Arjunolic acid 1MG
SDP
Catalog No. 502880222
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Arjunolic acid has antioxidant, anti-inflammatory, antinociceptive and anticholinesterasic (AChE and BuChE) activities, it may as promising targets for the development of innovative multi-functional medicines for Alzheimer desease treatment. Purity 98.64%
Catalog No. 50-288-0222 Supplier TARGETMOL CHEMICALS INC Supplier No. TN14031MG
Add to Cart
Add to Cart
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.Arjunolic acid has antioxidant, anti-inflammatory, antinociceptive and anticholinesterasic (AChE and BuChE) activities, it may as promising targets for the development of innovative multi-functional medicines for Alzheimer desease treatment. Purity 98.64%