Learn More
Cayman Chemical βARK1 Inhibitor, Purity: ≥95% (mixture of isomers), CAS: 24269-96-3, 5 mg
SDP
Catalog No. 502963920
Encompass_Preferred
An inhibitor of GRK2/βARK1 (IC50 = 126 µM); selective for GRK2/βARK1 over PKA at concentrations up to 1 mM; decreases systolic blood pressure in ob/ob and nicotinamide plus streptozotocin-induced mouse models of type 2 diabetes at 200 µg/kg; improves clonidine-induced relaxation of precontracted isolated aortic rings and improves glucose tolerance in the ob/ob mouse model; inhibits serotonin- or neurotensin-induced DIR in the rat ventral tegmental area in vitro
Catalog No. 50-296-3920 Supplier Cayman Chemical Supplier No. 217515 MG
An inhibitor of GRK2/βARK1 (IC50 = 126 µM); selective for GRK2/βARK1 over PKA at concentrations up to 1 mM; decreases systolic blood pressure in ob/ob and nicotinamide plus streptozotocin-induced mouse models of type 2 diabetes at 200 µg/kg; improves clonidine-induced relaxation of precontracted isolated aortic rings and improves glucose tolerance in the ob/ob mouse model; inhibits serotonin- or neurotensin-induced DIR in the rat ventral tegmental area in vitro