Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arkray™ DiaScreen™ 50 Urine Analyzer
SDP

Catalog No. 02675272
Change view
Click to view available options
Quantity:
50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-675-272 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-675-272 Supplier Arkray™ Supplier No. D10000

missing translation for 'validMainframeMsg'

  • For use with DiaScreen reagent strips for urinalysis with 3 or more analytes per strip
  • Requires only 5 seconds of technician time per test
  • Throughput of 50 tests per hour
  • Memory capacity of 200 most recent test results
  • Minimizes variability between operators
  • Patient identification number entered with standard keypad or with a barcode reader or keyboard
  • May interface with computer systems
  • Easy-to-read printouts flag abnormal test results with an asterisk
  • CLIA waived

Specifications

Product Type Diascreen 50 Urine Chemistry Analyzer
Humidity 20 to 85%
Memory 200 results
Temperature (English) Operating 59°F to 85°F
Temperature (Metric) Operating 15°C to 30°C
Depth (English) 12.83 in.
Depth (Metric) 32.5 cm
Height (English) 4.72 in.
Height (Metric) 11.9 cm
Width (English) 10.63 in.
Width (Metric) 27 cm
Weight (English) 4.4 lb.
Weight (Metric) 2 kg
Quantity 50
Amperage 1.2 A
Voltage 120/230 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.