Learn More
Arkray™ NicSafe™ Needle Incineration Replacement Cartridges
SDP
Catalog No. 02065187
Change view
Click to view available options
Replacement Cartridge; 2/pk
Catalog No.
02-065-187 Replacement Cartridge; 2/pk
1 options
Catalog No. 02-065-187 Supplier Arkray™ Supplier No. C500A07
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Replacement cartridges for use with NicSafe™ instant on-site incineration device
  • Incineration device disposes of a full range of hypodermic syringe needles (19 to 28G) using safe low voltage
  • Resulting debris is captured in a closed cartridge and discarded as biohazardous waste
  • Meets OSHA guidelines and has FDA 510(k) approval
NicSafe™ Needle Incineration Device:
  • Can be plugged into any standard wall outlet
  • Dimensions: 11.1D × 5.1W × 4.3 in.H