Learn More
Aviva Systems Biology Corp ARL11 Antibody - middle region (ARP52609_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52609_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARL11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARL11. Alternate names: ARLTS1, FLJ33930, MGC17429ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60783