Learn More
Aviva Systems Biology Corp ARL13B Antibody - middle region (ARP52779_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52779_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARL13B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARL13B. Alternate names: ARL2L1, DKFZp686E2075, DKFZp686L2472, DKFZp686M2074, DKFZp761H079, MGC120611, MGC120612, JBTS8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53699