Learn More
Aviva Systems Biology Corp Arl2bp Antibody - C-terminal region (ARP62205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Arl2bp. It was validated on Western Blot. . Alternate names: 1700010P10Rik, 1700027H16Rik, 6330544B05Rik, AI482273, AI849834, BART, Bart1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-610