Learn More
Aviva Systems Biology Corp ARL5A Antibody - middle region (ARP55757_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55757_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARL5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARL5A. Alternate names: ARFLP5, ARL5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-61601