Learn More
Medchemexpress LLC ARL67156 triethylamine | 00-00-0 | 99.9% | 1154.23 g/mol | C15H24Br2N5O12P3 · (4.3 C6H15N) | 25 MG
SDP
Catalog No. 5000246035
Encompass_Preferred
ARL67156 triethylamine is a small-molecule research reagent supplied as a high-purity solid for use as an NTPDase inhibitor in biochemical and cellular studies. It is provided in small-scale packages suitable for assay development and laboratory research.
  • High purity of approximately 99.9% for reliable experimental results.
  • Solid form with sealed storage to minimize moisture exposure.
  • Available in small milligram quantities for research workflows.
  • Reported to inhibit NTPDase enzymes for use in biochemical studies.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-46035 Supplier Medchemexpress LLC Supplier No. HY103265D25MG
ARL67156 triethylamine is a small-molecule research reagent supplied as a high-purity solid for use as an NTPDase inhibitor in biochemical and cellular studies. It is provided in small-scale packages suitable for assay development and laboratory research.
  • High purity of approximately 99.9% for reliable experimental results.
  • Solid form with sealed storage to minimize moisture exposure.
  • Available in small milligram quantities for research workflows.
  • Reported to inhibit NTPDase enzymes for use in biochemical studies.
  • Recommended storage: 4°C sealed; in solvent: -80°C (6 months) or -20°C (1 month).