Learn More
Medchemexpress LLC ARL67156 triethylamine | 99.9% | 1154.23 g/mol | C15H24Br2N5O12P3 · (4.3C6H15N) | 50 MG
SDP
Catalog No. 5000246004
Encompass_Preferred
ARL67156 triethylamine is a small-molecule ectonucleotidase inhibitor used in biochemical and pharmacological research to reduce extracellular ATP hydrolysis and downstream adenosine production. Supplied as a solid with high reported purity, it is intended for laboratory research use only.
  • Inhibits ecto-ATPase (CD39/CD73) activity in vitro and in vivo studies.
  • White to off-white solid suitable for laboratory use.
  • High purity (99.86%) as reported by the manufacturer.
  • Available in small lab pack sizes such as 50 MG solid and solution formats.
  • Store sealed at 4°C, protected from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-46004 Supplier Medchemexpress LLC Supplier No. HY103265D50MG
ARL67156 triethylamine is a small-molecule ectonucleotidase inhibitor used in biochemical and pharmacological research to reduce extracellular ATP hydrolysis and downstream adenosine production. Supplied as a solid with high reported purity, it is intended for laboratory research use only.
  • Inhibits ecto-ATPase (CD39/CD73) activity in vitro and in vivo studies.
  • White to off-white solid suitable for laboratory use.
  • High purity (99.86%) as reported by the manufacturer.
  • Available in small lab pack sizes such as 50 MG solid and solution formats.
  • Store sealed at 4°C, protected from moisture; in solvent: -80°C (6 months), -20°C (1 month).
  • For research use only; not for human or veterinary use.