Learn More
Medchemexpress LLC Arl67156 trisodium | 1021868-83-6 | 99.9% | 785.05 g·mol-1 | C15H21Br2N5Na3O12P3 | 1 MG
SDP
Catalog No. 5000245927
Encompass_Preferred
ARL67156 trisodium is a small-molecule research inhibitor of ecto-ATPase (NTPDase) enzymes, provided as the trisodium salt in solid form. It is used to study nucleotide hydrolysis and related pathologies such as calcific aortic valve disease and asthma.
  • Selective inhibitor of NTPDase1 (CD39), NTPDase3, and NPP1.
  • Reported Ki values approximately 11 μM (NTPDase1), 18 μM (NTPDase3), and 12 μM (NPP1).
  • High reported purity (99.94%).
  • Molecular weight about 785.05 g·mol-1 and exact formula C15H21Br2N5Na3O12P3.
  • Supplied as a solid trisodium salt, available in small research pack sizes.
  • Store sealed and away from moisture at -20°C; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-45927 Supplier Medchemexpress LLC Supplier No. HY1032651MG
ARL67156 trisodium is a small-molecule research inhibitor of ecto-ATPase (NTPDase) enzymes, provided as the trisodium salt in solid form. It is used to study nucleotide hydrolysis and related pathologies such as calcific aortic valve disease and asthma.
  • Selective inhibitor of NTPDase1 (CD39), NTPDase3, and NPP1.
  • Reported Ki values approximately 11 μM (NTPDase1), 18 μM (NTPDase3), and 12 μM (NPP1).
  • High reported purity (99.94%).
  • Molecular weight about 785.05 g·mol-1 and exact formula C15H21Br2N5Na3O12P3.
  • Supplied as a solid trisodium salt, available in small research pack sizes.
  • Store sealed and away from moisture at -20°C; in solution, store at -80°C (up to 6 months) or -20°C (up to 1 month).