Learn More
Medchemexpress LLC Arl67156 trisodium | 1021868-83-6 | MFCD08544552 | 99.9% | 785.05 g/mol | C15H21Br2N5Na3O12P3 | 5 MG
SDP
Catalog No. 5000247574
Encompass_Preferred
ARL67156 trisodium is the trisodium salt of ARL67156, a small-molecule inhibitor of ecto-ATPases (NTPDases) including CD39, NTPDase3, and NPP1. It is used in purinergic signaling research and disease models such as calcific aortic valve disease and asthma.
  • selective ecto-ATPase inhibitor targeting CD39, NTPDase3, and NPP1.
  • reported Ki values of ~11 μM, 18 μM, and 12 μM for respective targets.
  • high purity suitable for research (99.94% by HPLC).
  • small-scale research quantity (5 mg) for exploratory studies.
  • storage guidelines: sealed and dry; in solvent store at -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-47574 Supplier Medchemexpress LLC Supplier No. HY1032655MG
ARL67156 trisodium is the trisodium salt of ARL67156, a small-molecule inhibitor of ecto-ATPases (NTPDases) including CD39, NTPDase3, and NPP1. It is used in purinergic signaling research and disease models such as calcific aortic valve disease and asthma.
  • selective ecto-ATPase inhibitor targeting CD39, NTPDase3, and NPP1.
  • reported Ki values of ~11 μM, 18 μM, and 12 μM for respective targets.
  • high purity suitable for research (99.94% by HPLC).
  • small-scale research quantity (5 mg) for exploratory studies.
  • storage guidelines: sealed and dry; in solvent store at -80°C up to 6 months, -20°C up to 1 month.