Learn More
Medchemexpress LLC Arl67156 trisodium hydrate | 99.9% | 834.61 | C15H23Br2N5Na3O13P3 | 5 MG
SDP
Catalog No. 5000245968
Encompass_Preferred
ARL67156 trisodium hydrate is the trisodium hydrate salt form of ARL67156, a selective small-molecule inhibitor of ecto-ATPase enzymes including NTPDase1 (CD39), NTPDase3, and NPP1. Supplied as a white to off-white solid, it is intended for research use in studies of purinergic signaling, inflammation, and cardiovascular or respiratory disease models.
  • Selective inhibitor of ecto-ATPase enzymes, including NTPDase1 (CD39), NTPDase3, and NPP1.
  • Reported Kis values in the low micromolar range for primary targets.
  • Demonstrated activity in cellular and tissue assays such as potentiation of neurogenic contractions and modulation of neurotransmitter release.
  • High reported purity (99.94%), suitable for biochemical and cellular research.
  • Supplied as a solid, white to off-white, available in small research quantities.
Catalog No. 50-002-45968 Supplier Medchemexpress LLC Supplier No. HY103265B5MG
ARL67156 trisodium hydrate is the trisodium hydrate salt form of ARL67156, a selective small-molecule inhibitor of ecto-ATPase enzymes including NTPDase1 (CD39), NTPDase3, and NPP1. Supplied as a white to off-white solid, it is intended for research use in studies of purinergic signaling, inflammation, and cardiovascular or respiratory disease models.
  • Selective inhibitor of ecto-ATPase enzymes, including NTPDase1 (CD39), NTPDase3, and NPP1.
  • Reported Kis values in the low micromolar range for primary targets.
  • Demonstrated activity in cellular and tissue assays such as potentiation of neurogenic contractions and modulation of neurotransmitter release.
  • High reported purity (99.94%), suitable for biochemical and cellular research.
  • Supplied as a solid, white to off-white, available in small research quantities.