Learn More
Medchemexpress LLC ARL67156 trisodium hydrate | MFCD12910436 | 99.9% | 834.61 g/mol | C15H23Br2N5Na3O13P3 | 1 MG
SDP
Catalog No. 5000246263
Encompass_Preferred
ARL67156 trisodium hydrate is the trisodium salt hydrate form of ARL67156 (FPL 67156), a small-molecule inhibitor of ecto-ATPases used in research to study extracellular ATP signaling and disease models such as calcific aortic valve disease and asthma.
  • Inhibits ecto-ATPase enzymes, including NTPDase1 (CD39), NTPDase3, and NPP1.
  • Reported Ki values: 11 μM, 18 μM, and 12 μM for NTPDase1, NTPDase3, and NPP1, respectively.
  • Purity 99.94% as reported on the product page.
  • Supplied as a trisodium hydrate salt.
  • Available in small research quantities (for example, 1 mg and 5 mg sizes).
Catalog No. 50-002-46263 Supplier Medchemexpress LLC Supplier No. HY103265B1MG
ARL67156 trisodium hydrate is the trisodium salt hydrate form of ARL67156 (FPL 67156), a small-molecule inhibitor of ecto-ATPases used in research to study extracellular ATP signaling and disease models such as calcific aortic valve disease and asthma.
  • Inhibits ecto-ATPase enzymes, including NTPDase1 (CD39), NTPDase3, and NPP1.
  • Reported Ki values: 11 μM, 18 μM, and 12 μM for NTPDase1, NTPDase3, and NPP1, respectively.
  • Purity 99.94% as reported on the product page.
  • Supplied as a trisodium hydrate salt.
  • Available in small research quantities (for example, 1 mg and 5 mg sizes).