Learn More
Aviva Systems Biology Corp ARL8A Antibody - middle region (ARP52641_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52641_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARL8A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARL8A. Alternate names: ARL10B, FLJ45195, GIE2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-60793