Learn More
Aviva Systems Biology Corp ARL8B Antibody - middle region (ARP57138_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57138_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARL8B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARL8B. Alternate names: ARL10C, FLJ10702, Gie1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54036