Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific ASI ASO Latex Test Kit
SDP

Catalog No. 22415101 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-101 100 Tests
22-415-100 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-415-101 Supplier Arlington Scientific Supplier No. 300100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Two liquid controls

The ASI ASO Latex Test is a latex slide agglutination assay for the qualitative and semiquantitative detection of Antistreptolysin-O (ASO) antibodies in human serum.

Testing of Antistreptolysin-O (ASO) will detect the presence of antibodies from a prior Strep A infection. This is important in the diagnosis of post-infection complications of some heart and kidney diseases (Rheumatic fever and Glomerulonephritis). In the presence of antistreptolysin-O levels greater than 200 IU/mL in serum, coated latex particles will agglutinate providing a visual observable reaction of antigen and antibody.

  • Two-minute slide test
  • Results on AAB, CAP and API surveys
  • No initial dilution of patient samples is required for this test
  • ASO latex reagent consists of latex particles coated with streptolysin-O
  • Sensitivity: 200 IU/mL
  • Specificity: >95%

These materials are intended to be acquired, possessed, and used only by health professionals.

Specifications

CPT Code 86060
CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Detectable Analytes Anti-Streptolysin O (ASO)
Type ASO Latex Test Kit
Disposable Single-use
Includes Liquid control
Quantity 100 Tests
Sterility Non-sterile
Test Time 2 min.
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.