Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific ASI CRP Latex Test Kit
SDP

Catalog No. 22415112 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-112 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-112 Supplier Arlington Scientific Supplier No. 500100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ASI CRP Slide Test is a convenient and specific slide agglutination assay for the qualitative and semiquantitative detection of C-Reactive Protein (CRP) in human serum.

  • Two-minute slide test
  • Sensitivity: 6 mg/L
  • No initial dilution required
  • Two liquid controls included
  • Expires 24 months from date of manufacture
  • Results on AAB and other surveys

Specifications

CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Detectable Analytes CRP
Type CRP Latex Test Kit
Disposable Single-use
Includes Two liquid controls
Quantity 100 Tests
Test Time 2 min.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.