Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific RF Direct Rapid Latex Test
SDP

Catalog No. 22415114 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-114 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-114 Supplier Arlington Scientific Supplier No. 700100DC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detects Rheumatoid Factor associated with Rheumatoid Arthritis. Convenient and specific agglutination assay for qualitative and semiquantitative determination.

  • Latex agglutination test
  • Standardized to WHO reference
  • Two-minute slide test
  • Sensitivity: 8.0IU/mL
  • Specificity: >95%
  • No initial dilution required
  • NCCLS procedure sheets available
  • Results on AAB and other surveys

Specifications

Type RF Direct Slide Test
Includes Two liquid control
Quantity 100 Tests
Test Time 2 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.