Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific ASI RPR Carbon Antigen
SDP

Catalog No. 22415122 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-122 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-122 Supplier Arlington Scientific Supplier No. 901010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ASI’s non-treponemal RPR antigen screens for syphilis; a sexually transmitted disease caused by the bacterium Treponema pallidum (a spirochete bacteria). The disease is found worldwide and is easily treated if detected early. This test is FDA cleared for use in diagnostic screening.

For use with Arlington Scientific RPR Test Kits.

Specifications

CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Type RPR Carbon Antigen Suspension
Disposable Single-use
For Use With (Application) For ASI RPR Test Kits
Quantity 10 mL
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.