Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific ASI RPR Controls
SDP

Catalog No. p-2827417 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
3 x 2.5 mL
3 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-124 3 x 2.5 mL
22-415-125 3 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-415-124 Supplier Arlington Scientific Supplier No. 9050025
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

The ASI RPR Control Set is used with RPR and VDRL test kits to ensure proper performance. Contains Reacitve, Weak Reactive, and Nonreactive controls.

For use with RPR VDRL control serum sets.

Specifications

CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Type RPR/VDRL Control Set
Disposable Single-use
For Use With (Application) RPR Test Kit for Syphilis
Includes Reactive, weak reactive, nonreactive, CLSI package insert
Quantity 3 x 2.5 mL
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.