Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific RPR Test Kit
SDP

Catalog No. 22415121 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
500 Tests
5000 Test 10-well Card
5000 Test 30-well Card
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-121 5000 Test 30-well Card
22-415-116 100 Tests
22-415-119 500 Tests
22-415-120 5000 Test 10-well Card
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 22-415-121 Supplier Arlington Scientific Supplier No. 900500030
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Qualitative and semiquantitative Syphilis screening test

  • Available with or without controls
  • Carbon antigen suspension
  • Easy to interpret
  • Sensitivity: >98%
  • Specificity: >99%
  • Color-coded, screw cap dropper vials—no glass ampules
  • Liquid controls
  • Three levels: reactive, weak reactive, and nonreactive
  • Results on CAP and AAB surveys
  • All testing materials provided
  • NCCLS procedure sheets available
  • CPD, CPDA-1 and EDTA may be used

Specifications

Detectable Analytes Reagin (Syphilis)
Type RPR Test Card
For Use With (Application) Syphilis Testing
Includes RPR Carbon Antigen 3 levels of Serum control 18mm circle cards spreading pipets Antigen dispensing bottle and needle, thirty-well cards
Quantity 5000 Test 30-well Card
Specificity >99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.