Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arlington Scientific ASI SLE Test Kit
SDP

Catalog No. 22415105 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-415-105 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-105 Supplier Arlington Scientific Supplier No. 350050
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ASI SLE Slide Test is a convenient and specific latex slide agglutination assay for the qualitative and semiquantitative detection of anti-deoxyribonucleoprotein (anti-DNP) in human serum.

  • One-minute test
  • Accurate screening test for SLE
  • Sensitivity: >86%
  • Specificity: >82%
  • Two liquid controls included
  • No initial dilution required

These materials are intended to be acquired, possessed, and used only by health professionals.

Specifications

Detectable Analytes Anti-DNP Antibodies
Type SLE Test Kit
Disposable Single-use
Includes Two liquid control, NCCLS procedure sheet
Quantity 50 Tests
Test Time 1 min.
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.