Learn More
Arlington Scientific ASI Staph Test Kit
SDP
Catalog No. 22415104 Shop All Arlington Scientific Products
Change view
Click to view available options
Quantity:
50 Tests
Catalog No. Quantity
22-415-104 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. 22-415-104 Supplier Arlington Scientific Supplier No. 280250C
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
The ASI Color Staph is a rapid, color enhanced latex agglutination test to detect clumping factor and/or Protein-A, two characteristics associated with Staphylococcus aureus colonies obtained from culture.
  • CLIA Complexity: High
  • Latex agglutination test
  • 45-second slide test
  • Greater than 99% correlation with tube coagulase test
  • Simple to interpret
  • Expiry 12 months from date of manufacture
Specifications
CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Detectable Analytes S. aureus Coagulase, S. aureus Coagulase, Protein A
Type Staph Test Kit
Disposable Single-use
Includes Liquid control
Quantity 50 Tests
Form Ready-to-Use (RTU)