Learn More
Bio X Cell IgG Isotype Control, Armenian Hamster, Anti-S. Japonicum, Purified, 5 MG
Supplier: Bio X Cell BE0260R005MG
This Armenian hamster IgG isotype control antibody reacts with glutathione-S-transferase (GST) of *Schistosoma japonicum* origin. It serves as an excellent isotype control for Armenian hamster IgG antibodies in most in vitro and in vivo experiments, as mammals do not express *S. japonicum* GST (SjGST). Additionally, it facilitates the detection, purification, and quantification of SjGST-tagged recombinant proteins.
- Monoclonal antibody, clone PIP.
- Reacts with *Schistosoma japonicum* glutathione-S-transferase (GST).
- Functions as an isotype control for Armenian hamster IgG antibodies in in vitro and in vivo experiments.
- Aids in the detection, purification, and quantification of SjGST-tagged recombinant proteins.
- Produced in an animal-free facility.
- Purified using Protein A.
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives.
- Endotoxin level: 1 EU/mg.
- Purity: 95%, as determined by S
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.