Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Armadillo™ Low-Profile PCR Strip Plate, 96 well, green

Catalog No. FERAB2696G Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERAB2696G 25 plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERAB2696G Supplier Thermo Scientific™ Supplier No. AB2696G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific Armadillo 96-well low-profile PCR strip plates are the ultimate in high-performance PCR plastics. They combine the rigidity of a polycarbonate frame with thin-walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.

  • Armadillo 96-well low-profile PCR strip plate combine the rigidity of a polycarbonate frame, with thin-walled polypropylene wells
  • Plates provide superior thermal cycling performance under all conditions without warping
  • Thermostable polycarbonate semi-skirted frame design for warp-resistance during thermal cycling and enhanced mechanical stability during robotic handling
  • Breakable into 8-tube strips to accommodate variable numbers of samples and minimize waste
  • Optically clear skirt aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • ANSI:SBS footprint ensures complete compatibility with robotic systems for high-throughput applications
  • Thin-walled U-bottom wells for optimal heat transfer and maximum sample recovery
  • Black alpha-numeric lettering for easy well identification
  • Integrated frame and well design for improved heat sealing
  • Low profile design for maximum heat transfer and minimal evaporation with low-volume samples (200 μL working volume)
  • Compatible with Roche LightCycler™ 480 Real-Time PCR systems
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Breakapart Yes
Array 8 x 12
Packaging 25 Plates
Product Line Armadillo
Color Green
Product Type PCR Microplate
For Use With (Equipment) Real-Time PCR Systems or Thermal Cycler
Skirt Style Semi
Material Polycarbonate
Volume (Metric) Well 0.2 mL
No. of Wells 96
Volume (Metric) Working 0.1 mL
Well Shape Round Bottom (U-well)
Quantity 25 plates
Reaction Speed Low Profile
Well Color Clear
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.