Learn More
Aviva Systems Biology Corp ARMC3 Antibody - middle region (ARP55626_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55626_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARMC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ARMC3. Alternate names: FLJ25845, FLJ32827, RP11-300B11.1, CT81, KU-CT-1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-61559