Learn More
Aviva Systems Biology Corp ARMC4 Antibody - N-terminal region (ARP58586_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58586_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARMC4. It was validated on Western Blot. . Alternate names: DKFZp434P1735, FLJ10376, FLJ10817, FLJ32798, RP11-691I13.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Rat
Catalog No. 50-286-487