Learn More
Aviva Systems Biology Corp ARMC8 Antibody - N-terminal region (ARP56019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARMC8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ARMC8. Alternate names: HSPC056, MGC10058, MGC4880, S863-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-286-110