Learn More
Aviva Systems Biology Corp ARMCX1 Antibody - C-terminal region (ARP47286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARMCX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ARMCX1. Alternate names: ALEX1, DKFZp686P06199ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59508