Learn More
Rockland Immunochemicals Armenian Hamster IgG (H&L) Antibody 2 mg
SDP
Supplier:  Rockland Immunochemicals 619101002
Encompass
Anti-Armenian Hamster IgG Antibody generated in goat detects Armenian Hamster IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3942