Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ARMET/MANF Human H 10ug
SDP

Catalog No. 5000195847
Encompass_Preferred

Recombinant human ARMET/MANF (His-tagged) is supplied lyophilized and intended for research applications involving neuronal survival and cellular homeostasis. The product is expressed in Escherichia coli, formulated in a Tris-NaCl buffer, and characterized by SDS-PAGE for purity and molecular weight. Store lyophilized at -20°C and aliquot after reconstitution for prolonged storage.

  • Recombinant human protein for research use.
  • N-terminal hexahistidine tag for affinity purification.
  • Expressed in Escherichia coli and lyophilized for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Observed molecular weight approximately 20 kDa under reducing conditions.
  • Formulated in Tris-NaCl and 0.22 μm filtered prior to lyophilization.

Catalog No. 50-001-95847 Supplier Medchemexpress LLC Supplier No. HYP71940A10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human ARMET/MANF (His-tagged) is supplied lyophilized and intended for research applications involving neuronal survival and cellular homeostasis. The product is expressed in Escherichia coli, formulated in a Tris-NaCl buffer, and characterized by SDS-PAGE for purity and molecular weight. Store lyophilized at -20°C and aliquot after reconstitution for prolonged storage.

  • Recombinant human protein for research use.
  • N-terminal hexahistidine tag for affinity purification.
  • Expressed in Escherichia coli and lyophilized for stability.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Observed molecular weight approximately 20 kDa under reducing conditions.
  • Formulated in Tris-NaCl and 0.22 μm filtered prior to lyophilization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.