Learn More
Medchemexpress LLC ARMET/MANF Human H 2ug
SDP
Catalog No. 5000196411
Encompass_Preferred
Recombinant human ARMET/MANF protein with a histidine tag, supplied as lyophilized powder for research applications. The protein has an approximate molecular weight of 20 kDa and is provided at high purity (≥95%), suitable for biochemical and cell biology assays. Follow recommended reconstitution and storage instructions to maintain activity and longevity.
  • His-tagged recombinant human protein for affinity purification and detection.
  • High purity (≥95%) verified by reducing SDS-PAGE.
  • Approximate molecular weight of 20 kDa for assay planning.
  • Supplied as lyophilized powder for extended shelf life.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O to minimize material loss.
  • Stable when stored at -20°C; aliquot and add carrier protein for long-term storage.
Catalog No. 50-001-96411 Supplier Medchemexpress LLC Supplier No. HYP71940A2UG
Recombinant human ARMET/MANF protein with a histidine tag, supplied as lyophilized powder for research applications. The protein has an approximate molecular weight of 20 kDa and is provided at high purity (≥95%), suitable for biochemical and cell biology assays. Follow recommended reconstitution and storage instructions to maintain activity and longevity.
  • His-tagged recombinant human protein for affinity purification and detection.
  • High purity (≥95%) verified by reducing SDS-PAGE.
  • Approximate molecular weight of 20 kDa for assay planning.
  • Supplied as lyophilized powder for extended shelf life.
  • Recommended reconstitution at ≥100 μg/mL in ddH2O to minimize material loss.
  • Stable when stored at -20°C; aliquot and add carrier protein for long-term storage.