Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ARMET/MANF Human H 500ug
SDP

Catalog No. 5000196131
Encompass_Preferred

Recombinant human mesencephalic astrocyte-derived neurotrophic factor (MANF) supplied lyophilized with an N-terminal His tag. The product corresponds to UniProt P55145 (residues L25-L182), is expressed in E. coli, and is provided at high purity for research applications including studies of neuroprotection, endoplasmic reticulum stress, and cell signaling.

  • His-tagged for affinity purification and detection.
  • Expressed in E. coli for efficient production.
  • Contains residues L25-L182 of the human sequence.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Lyophilized for long-term storage; aliquot and freeze for extended stability.
  • Suitable for in vitro research on neuroprotection and ER stress pathways.

Catalog No. 50-001-96131 Supplier Medchemexpress LLC Supplier No. HYP71940A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human mesencephalic astrocyte-derived neurotrophic factor (MANF) supplied lyophilized with an N-terminal His tag. The product corresponds to UniProt P55145 (residues L25-L182), is expressed in E. coli, and is provided at high purity for research applications including studies of neuroprotection, endoplasmic reticulum stress, and cell signaling.

  • His-tagged for affinity purification and detection.
  • Expressed in E. coli for efficient production.
  • Contains residues L25-L182 of the human sequence.
  • Purity ≥95% as determined by reducing SDS-PAGE.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Lyophilized for long-term storage; aliquot and freeze for extended stability.
  • Suitable for in vitro research on neuroprotection and ER stress pathways.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.