Learn More
Medchemexpress LLC ARMET/MANF Mouse H 1mg | 1mg
SDP
Catalog No. 5000186542
Encompass_Preferred
ARMET/MANF proteins regulate the survival of dopaminergic neurons in the ventral midbrain and modulate GABAergic transmission to dopaminergic neurons in the substantia nigra It enhances GABA inhibitory currents and inhibits cell proliferation while preventing ER stress-induced cell death ARMET/MANF Protein Mouse (HEK293 His) is the recombinant mouse-derived ARMET/MANF protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-86542 Supplier Medchemexpress LLC Supplier No. HYP772961MG
ARMET/MANF proteins regulate the survival of dopaminergic neurons in the ventral midbrain and modulate GABAergic transmission to dopaminergic neurons in the substantia nigra It enhances GABA inhibitory currents and inhibits cell proliferation while preventing ER stress-induced cell death ARMET/MANF Protein Mouse (HEK293 His) is the recombinant mouse-derived ARMET/MANF protein expressed by HEK293 with C-His labeled tag