Learn More
Sino Biological ARMET/MANF Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11324T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ARMET/MANF (rh ARMET/MANF; Catalog#11324-H08H; NP_006001.3; Met1-Leu182). ARMET/MANF specific IgG was purified by Human ARMET/MANF affinity chromatography.
Catalog No. 50-224-8612