Learn More
Medchemexpress LLC Armillarisin A | 53696-74-5 | MFCD01662561 | 99.8% | 234.20 g/mol | C12H10O5 | 1 ML
SDP
Catalog No. 5000241835
Encompass_Preferred
Armillarisin A is a research-grade small molecule (C12H10O5, CAS 53696-74-5) used as an analytical standard and biological probe. It has reported immunomodulatory effects - increasing IL-4 and decreasing IL-1β - and has been investigated in ulcerative colitis studies. The product is supplied as ready-to-use solutions and as solids for analytical and research applications.
Catalog No. 50-002-41835 Supplier Medchemexpress LLC Supplier No. HY10801310MM 1ML
Armillarisin A is a research-grade small molecule (C12H10O5, CAS 53696-74-5) used as an analytical standard and biological probe. It has reported immunomodulatory effects - increasing IL-4 and decreasing IL-1β - and has been investigated in ulcerative colitis studies. The product is supplied as ready-to-use solutions and as solids for analytical and research applications.