Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 3µm, 10cm x 3.0mm ID
Catalog No. 5000805454
Encompass
Armor C18 Analytical Column. 120Â, 3µm, 10cm x 3.0mm ID
Catalog No. 50-008-05454 Supplier Analytical Sales & Services Supplier No. ADV7305
Armor C18 Analytical Column. 120Â, 3µm, 10cm x 3.0mm ID