Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 3µm, 3cm x 2.1mm ID
Catalog No. 5000805751
Encompass
Armor C18 Analytical Column. 120Â, 3µm, 3cm x 2.1mm ID
Catalog No. 50-008-05751 Supplier Analytical Sales & Services Supplier No. ADV7321
Armor C18 Analytical Column. 120Â, 3µm, 3cm x 2.1mm ID