Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 3µm, 5cm x 4.6mm ID
Catalog No. 5000806173
Encompass
Armor C18 Analytical Column. 120Â, 3µm, 5cm x 4.6mm ID
Catalog No. 50-008-06173 Supplier Analytical Sales & Services Supplier No. ADV7300
Armor C18 Analytical Column. 120Â, 3µm, 5cm x 4.6mm ID