Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 5000806089
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID
Catalog No. 50-008-06089 Supplier Analytical Sales & Services Supplier No. ADV7009
Armor C18 Analytical Column. 120Â, 5µm, 10cm x 2.1mm ID