Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 5000805997
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID
Catalog No. 50-008-05997 Supplier Analytical Sales & Services Supplier No. ADV7001
Armor C18 Analytical Column. 120Â, 5µm, 10cm x 4.6mm ID