Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 25cm x 2.1mm ID
Catalog No. 5000806159
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 25cm x 2.1mm ID
Catalog No. 50-008-06159 Supplier Analytical Sales & Services Supplier No. ADV7011
Add to Cart
Add to Cart
Armor C18 Analytical Column. 120Â, 5µm, 25cm x 2.1mm ID