Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 5000806752
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID
Catalog No. 50-008-06752 Supplier Analytical Sales & Services Supplier No. ADV7007
Armor C18 Analytical Column. 120Â, 5µm, 25cm x 3.0mm ID