Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 3cm x 3.0mm ID
Catalog No. 5000806160
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 3cm x 3.0mm ID
Catalog No. 50-008-06160 Supplier Analytical Sales & Services Supplier No. ADV7030
Armor C18 Analytical Column. 120Â, 5µm, 3cm x 3.0mm ID