Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 5cm x 3.0mm ID
Catalog No. 5000805626
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 5cm x 3.0mm ID
Catalog No. 50-008-05626 Supplier Analytical Sales & Services Supplier No. ADV7004
Armor C18 Analytical Column. 120Â, 5µm, 5cm x 3.0mm ID