Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 5µm, 5cm x 4.6mm ID
Catalog No. 5000806661
Encompass
Armor C18 Analytical Column. 120Â, 5µm, 5cm x 4.6mm ID
Catalog No. 50-008-06661 Supplier Analytical Sales & Services Supplier No. ADV7000
Armor C18 Analytical Column. 120Â, 5µm, 5cm x 4.6mm ID