Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 3µm, 10cm x 0.3mm ID
Catalog No. 5000806907
Encompass
Armor C18 Microbore Column. 120Â, 3µm, 10cm x 0.3mm ID
Catalog No. 50-008-06907 Supplier Analytical Sales & Services Supplier No. ADV10M3A203
Armor C18 Microbore Column. 120Â, 3µm, 10cm x 0.3mm ID