Learn More
Analytical Sales & Services Armor C18 Microbore Column. 120Â, 3µm, 10cm x 1.0mm ID
Catalog No. 5000805658
Encompass
Armor C18 Microbore Column. 120Â, 3µm, 10cm x 1.0mm ID
Catalog No. 50-008-05658 Supplier Analytical Sales & Services Supplier No. ADV10M1A203
Armor C18 Microbore Column. 120Â, 3µm, 10cm x 1.0mm ID